siRNA Details
| Gene Symbol: Dlgap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008767428.1 | XP_008765650.1 | 6 | 2225 | ||
| XM_008767429.1 | XP_008765651.1 | 6 | 1425 | |||
| XM_008767430.1 | XP_008765652.1 | 6 | 1456 | |||
| XM_008767431.1 | XP_008765653.1 | 6 | 1450 | |||
| XM_008767432.1 | XP_008765654.1 | 6 | 1900 | |||
| XM_008767433.1 | XP_008765655.1 | 6 | 1112 | |||
| GenBank mRNA | AB003594.1 | - | 6 | 1098 | ||
| FQ212368.1 | - | 6 | 1214 | |||
| U67137.1 | - | 8 | 2245 | |||
| U67987.1 | - | 6 | 1039 | |||

Dlgap1
Genomic Map