siRNA Details
| Gene Symbol: Gapdhs | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023964.1 | NP_076454.1 | 4 | 468 | ||
| XM_006228790.2 | XP_006228852.1 | 4 | 838 | |||
| XM_006228791.2 | XP_006228853.1 | 4 | 699 | |||
| GenBank mRNA | AJ297631.1 | - | 4 | 468 | ||
| BC161847.1 | - | 4 | 490 | 7112898 | ||

Gapdhs
Genomic Map