siRNA Details
| Gene Symbol: Wdr7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023975.1 | NP_076465.1 | 4 | 449 | ||
| GenBank mRNA | AF192379.1 | - | 4 | 449 | ||
| AF305813.1 | - | 4 | 449 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Wdr7| Gene Symbol: Wdr7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023975.1 | NP_076465.1 | 4 | 449 | ||
| GenBank mRNA | AF192379.1 | - | 4 | 449 | ||
| AF305813.1 | - | 4 | 449 | |||