siRNA Details
| Gene Symbol: Apaf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023979.1 | NP_076469.1 | 2 | 742 | ||
| GenBank mRNA | AF218388.1 | - | 2 | 197 | ||
| AF320222.1 | - | 2 | 742 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Apaf1| Gene Symbol: Apaf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023979.1 | NP_076469.1 | 2 | 742 | ||
| GenBank mRNA | AF218388.1 | - | 2 | 197 | ||
| AF320222.1 | - | 2 | 742 | |||