siRNA Details
| Gene Symbol: Mepe | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024142.1 | NP_077056.1 | 3 | 160 | ||
| XM_006250634.2 | XP_006250696.1 | 2 | 228 | |||
| XM_006250635.2 | XP_006250697.1 | 2 | 1794 | |||
| GenBank mRNA | AF260922.1 | - | 3 | 160 | ||
| AF530558.1 | - | 3 | 157 | |||
| AF530559.1 | - | 2 | 136 | |||

Mepe
Genomic Map