siRNA Details
| Gene Symbol: Rassf9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022959.2 | NP_075248.1 | 2 | 462 | ||
| GenBank mRNA | AF056208.1 | - | 2 | 462 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rassf9| Gene Symbol: Rassf9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022959.2 | NP_075248.1 | 2 | 462 | ||
| GenBank mRNA | AF056208.1 | - | 2 | 462 | ||