siRNA Details
| Gene Symbol: Gabre | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023091.1 | NP_075579.1 | 4 | 1965 | ||
| GenBank mRNA | AF189262.1 | - | 4 | 1965 | ||
| AF255385.1 | - | 1527 | ||||
| AF255386.1 | - | 2 | 220 | |||
| AF255387.1 | - | 2 | 220 | |||
| AF255612.1 | - | 530 | ||||

Gabre
Genomic Map