siRNA Details
| Gene Symbol: Mgat5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023095.1 | NP_075583.1 | 3 | 482 | ||
| GenBank mRNA | L14284.1 | - | 3 | 482 | ||
| Gene Symbol: LOC100909582 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006249676.2 | XP_006249738.1 | 3 | 834 | ||
| XM_006249677.2 | XP_006249739.1 | 3 | 831 | |||
| XM_006249679.2 | XP_006249741.1 | 2 | 406 | |||
| XM_008769509.1 | XP_008767731.1 | 4 | 982 | |||

Mgat5
Genomic Map