siRNA Details
| Gene Symbol: Cd200r1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023953.1 | NP_076443.1 | 3 | 277 | ||
| XM_006248347.2 | XP_006248409.1 | 3 | 590 | |||
| XM_008768743.1 | XP_008766965.1 | 3 | 577 | |||
| GenBank mRNA | AY583211.1 | - | 3 | 277 | ||
| Gene Symbol: Cd200r1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008768755.1 | XP_008766977.1 | 5 | 578 | ||
| XM_008768756.1 | XP_008766978.1 | 5 | 534 | |||
| XM_008768757.1 | XP_008766979.1 | 4 | 524 | |||
| XM_008768758.1 | XP_008766980.1 | 4 | 532 | |||

Cd200r1
Genomic Map