siRNA Details
Gene Symbol: Cdx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023963.1 | NP_076453.1 | 2 | 723 | ||
XM_008768999.1 | XP_008767221.1 | 2 | 614 | |||
GenBank mRNA | AJ278466.1 | - | 2 | 723 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Cdx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023963.1 | NP_076453.1 | 2 | 723 | ||
XM_008768999.1 | XP_008767221.1 | 2 | 614 | |||
GenBank mRNA | AJ278466.1 | - | 2 | 723 |