siRNA Details
| Gene Symbol: DUSP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004418.3 | NP_004409.1 | 4 | 940 | ||
| XM_017003546.1 | XP_016859035.1 | 3 | 1052 | |||
| GenBank mRNA | AF331843.1 | - | 1 | 700 | ||
| BC007771.2 | - | 4 | 916 | 4297852 | ||
| L11329.1 | - | 4 | 939 | |||

DUSP2
Genomic Map