siRNA Details
| Gene Symbol: Fgfr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024146.1 | NP_077060.1 | 3 | 419 | ||
| XM_006253323.2 | XP_006253385.1 | 3 | 427 | |||
| XM_006253324.2 | XP_006253386.1 | 4 | 640 | |||
| XM_006253325.2 | XP_006253387.1 | 2 | 160 | |||
| XM_006253326.2 | XP_006253388.1 | 2 | 160 | |||
| XM_006253327.1 | XP_006253389.1 | 3 | 447 | |||
| GenBank mRNA | D12498.1 | - | 3 | 419 | ||
| S54008.1 | - | 3 | 452 | |||

Fgfr1
Genomic Map