siRNA Details
| Gene Symbol: Barhl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022956.2 | NP_075245.1 | 2 | 708 | ||
| GenBank mRNA | AB004056.1 | - | 2 | 712 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Barhl2| Gene Symbol: Barhl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022956.2 | NP_075245.1 | 2 | 708 | ||
| GenBank mRNA | AB004056.1 | - | 2 | 712 | ||