siRNA Details
| Gene Symbol: Csnk1g2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033870.1 | NP_001029042.1 | 12 | 1683 | ||
| NM_023102.2 | NP_075590.2 | 12 | 1680 | |||
| XM_006241000.2 | XP_006241062.1 | 11 | 3058 | |||
| XM_006241001.2 | XP_006241063.1 | 11 | 1766 | |||
| XM_006241002.2 | XP_006241064.1 | 12 | 2978 | |||
| XM_006241003.2 | XP_006241065.1 | 11 | 2408 | |||
| XM_008765130.1 | XP_008763352.1 | 11 | 1766 | |||
| GenBank mRNA | BC072533.1 | - | 12 | 1683 | 7098992 | |
| BC099758.1 | - | 6 | 542 | |||
| U22297.1 | - | 11 | 1414 | |||

Csnk1g2
Genomic Map