siRNA Details
Gene Symbol: Camkv | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024000.1 | NP_076490.1 | 6 | 607 | ||
GenBank mRNA | L22557.1 | - | 6 | 607 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Camkv | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024000.1 | NP_076490.1 | 6 | 607 | ||
GenBank mRNA | L22557.1 | - | 6 | 607 |