siRNA Details
| Gene Symbol: Mrs2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024001.1 | NP_076491.1 | 4 | 360 | ||
| XM_006253937.2 | XP_006253999.1 | 4 | 478 | |||
| GenBank mRNA | AF288289.1 | - | 4 | 360 | ||
| BC083554.1 | - | 4 | 336 | |||
| BC098916.1 | - | 4 | 335 | |||

Mrs2
Genomic Map