siRNA Details
| Gene Symbol: Hspa8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024351.2 | NP_077327.1 | 2 | 163 | ||
| GenBank mRNA | BC061547.1 | - | 2 | 170 | 6921606 | |
| BC098914.1 | - | 2 | 115 | 7104230 | ||
| M11942.1 | - | 2 | 134 | |||
| Gene Symbol: LOC689955 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008764620.1 | XP_008762842.1 | 1 | 165 | ||

Hspa8
Genomic Map