siRNA Details
Gene Symbol: Chrna4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024354.1 | NP_077330.1 | 5 | 443 | ||
GenBank mRNA | AF007212.1 | - | 5 | 443 | ||
L31620.1 | - | 444 | ||||
M15682.1 | - | 5 | 422 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Chrna4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024354.1 | NP_077330.1 | 5 | 443 | ||
GenBank mRNA | AF007212.1 | - | 5 | 443 | ||
L31620.1 | - | 444 | ||||
M15682.1 | - | 5 | 422 |