siRNA Details
| Gene Symbol: Frk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024368.1 | NP_077344.1 | 1 | 585 | ||
| GenBank mRNA | U09583.1 | - | 1 | 585 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Frk| Gene Symbol: Frk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024368.1 | NP_077344.1 | 1 | 585 | ||
| GenBank mRNA | U09583.1 | - | 1 | 585 | ||