siRNA Details
| Gene Symbol: Slc6a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024371.2 | NP_077347.1 | 2 | 256 | ||
| XM_006237059.1 | XP_006237121.1 | 3 | 465 | |||
| XM_006237060.2 | XP_006237122.1 | 3 | 2685 | |||
| GenBank mRNA | M59742.1 | - | 2 | 256 | ||

Slc6a1
Genomic Map