siRNA Details
Gene Symbol: Gucy2e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024380.1 | NP_077356.1 | 4 | 1313 | ||
GenBank mRNA | L36029.1 | - | 4 | 1313 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gucy2e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024380.1 | NP_077356.1 | 4 | 1313 | ||
GenBank mRNA | L36029.1 | - | 4 | 1313 |