siRNA Details
| Gene Symbol: Serpind1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024382.1 | NP_077358.1 | 3 | 191 | ||
| XM_006248709.2 | XP_006248771.1 | 2 | 198 | |||
| GenBank mRNA | BC091145.1 | - | 3 | 171 | 7372906 | |
| FQ218607.1 | - | 2 | 168 | |||
| X74549.1 | - | 3 | 191 | |||
| X74550.1 | - | 2 | 138 | |||

Serpind1
Genomic Map