siRNA Details
| Gene Symbol: HOXC6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004503.3 | NP_004494.1 | 2 | 1526 | ||
| NM_153693.4 | NP_710160.1 | 3 | 1923 | |||
| GenBank mRNA | M16938.1 | - | 3 | 1637 | ||
| S82986.1 | - | 2 | 1540 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

HOXC6| Gene Symbol: HOXC6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004503.3 | NP_004494.1 | 2 | 1526 | ||
| NM_153693.4 | NP_710160.1 | 3 | 1923 | |||
| GenBank mRNA | M16938.1 | - | 3 | 1637 | ||
| S82986.1 | - | 2 | 1540 | |||