siRNA Details
| Gene Symbol: Grb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030846.2 | NP_110473.2 | 2, 3 | 273 | ||
| GenBank mRNA | AA926364.1 | - | 2, 3 | 273 | ||
| BC091144.1 | - | 1, 2 | 217 | 7382476 | ||
| D49846.1 | - | 2, 3 | 388 | |||
| D49847.1 | - | 2, 3 | 230 | |||
| FQ220840.1 | - | 2, 3 | 339 | |||
| X62853.1 | - | 1, 2 | 123 | |||

Grb2
Genomic Map