siRNA Details
| Gene Symbol: Gabrg3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024370.3 | NP_077346.3 | 4, 5 | 619 | ||
| GenBank mRNA | M81142.1 | - | 4, 5 | 601 | ||
| X63324.1 | - | 675 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gabrg3| Gene Symbol: Gabrg3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024370.3 | NP_077346.3 | 4, 5 | 619 | ||
| GenBank mRNA | M81142.1 | - | 4, 5 | 601 | ||
| X63324.1 | - | 675 | ||||