siRNA Details
| Gene Symbol: Agxt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001276706.1 | NP_001263635.1 | 2 | 368 | ||
| NM_030656.2 | NP_085914.2 | 2 | 368 | |||
| GenBank mRNA | BC088133.1 | - | 2 | 200 | ||
| M35270.1 | - | 2 | 368 | |||
| X06357.1 | - | 2 | 306 | |||

Agxt
Genomic Map