siRNA Details
| Gene Symbol: Luzp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030830.2 | NP_110457.2 | 4 | 607 | ||
| XM_006239274.2 | XP_006239336.1 | 3 | 550 | |||
| GenBank mRNA | AF181259.1 | - | 5 | 2557 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Luzp1| Gene Symbol: Luzp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030830.2 | NP_110457.2 | 4 | 607 | ||
| XM_006239274.2 | XP_006239336.1 | 3 | 550 | |||
| GenBank mRNA | AF181259.1 | - | 5 | 2557 | ||