siRNA Details
| Gene Symbol: Grb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030846.2 | NP_110473.2 | 3 | 309 | ||
| GenBank mRNA | AA926364.1 | - | 3 | 309 | ||
| BC091144.1 | - | 2 | 253 | 7382476 | ||
| D49846.1 | - | 3 | 424 | |||
| D49847.1 | - | 3 | 266 | |||
| D49848.1 | - | 2 | 136 | |||
| FQ220840.1 | - | 3 | 375 | |||
| X62853.1 | - | 2 | 159 | |||

Grb2
Genomic Map