siRNA Details
| Gene Symbol: Asic1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024154.2 | NP_077068.1 | 12 | 1685 | ||
| XM_006257378.2 | XP_006257440.1 | 10 | 2010 | |||
| XM_006257381.2 | XP_006257443.1 | 12 | 1970 | |||
| XM_008765793.1 | XP_008764015.1 | 9 | 2147 | |||
| XM_008765794.1 | XP_008764016.1 | 11 | 2106 | |||
| XM_008765795.1 | XP_008764017.1 | 11 | 1759 | |||
| XM_008765796.1 | XP_008764018.1 | 11 | 2103 | |||
| GenBank mRNA | AB049451.1 | - | 10 | 1276 | ||
| AJ006519.1 | - | 10 | 1731 | |||
| AJ309926.1 | - | 10 | 1735 | |||
| CO398759.1 | - | 5 | 430 | |||

Asic1
Genomic Map