siRNA Details
| Gene Symbol: Grid2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024379.1 | NP_077355.1 | 4 | 865 | ||
| GenBank mRNA | U08256.1 | - | 4 | 865 | ||
| Z17239.1 | - | 4 | 578 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Grid2| Gene Symbol: Grid2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024379.1 | NP_077355.1 | 4 | 865 | ||
| GenBank mRNA | U08256.1 | - | 4 | 865 | ||
| Z17239.1 | - | 4 | 578 | |||