siRNA Details
Gene Symbol: Gucy2e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024380.1 | NP_077356.1 | 14, 15 | 2851 | ||
GenBank mRNA | L36029.1 | - | 14, 15 | 2851 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
