siRNA Details
| Gene Symbol: Aco2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024398.2 | NP_077374.2 | 4 | 456 | ||
| GenBank mRNA | AJ243266.1 | - | 4 | 457 | ||
| BC061999.1 | - | 4 | 456 | 5621226 | ||
| FQ216755.1 | - | 4 | 481 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Aco2| Gene Symbol: Aco2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024398.2 | NP_077374.2 | 4 | 456 | ||
| GenBank mRNA | AJ243266.1 | - | 4 | 457 | ||
| BC061999.1 | - | 4 | 456 | 5621226 | ||
| FQ216755.1 | - | 4 | 481 | |||