siRNA Details
| Gene Symbol: Akap4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024402.1 | NP_077378.1 | 5 | 491 | ||
| GenBank mRNA | AF008114.1 | - | 5 | 491 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Akap4| Gene Symbol: Akap4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024402.1 | NP_077378.1 | 5 | 491 | ||
| GenBank mRNA | AF008114.1 | - | 5 | 491 | ||