siRNA Details
| Gene Symbol: Acvr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024486.1 | NP_077812.1 | 4 | 595 | ||
| XM_006234214.2 | XP_006234276.1 | 6 | 2200 | |||
| XM_008761862.1 | XP_008760084.1 | 7 | 2257 | |||
| XM_008761863.1 | XP_008760085.1 | 7 | 4501 | |||
| XM_008761864.1 | XP_008760086.1 | 7 | 4247 | |||
| XM_008761865.1 | XP_008760087.1 | 7 | 4290 | |||
| XM_008761866.1 | XP_008760088.1 | 7 | 967 | |||
| XM_008761867.1 | XP_008760089.1 | 4 | 511 | |||
| GenBank mRNA | BC167754.1 | - | 5 | 766 | 9089038 | |
| L19341.1 | - | 4 | 595 | |||

Acvr1
Genomic Map