siRNA Details
| Gene Symbol: Fabp7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030832.2 | NP_110459.1 | 3 | 419 | ||
| GenBank mRNA | U02096.1 | - | 3 | 420 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fabp7| Gene Symbol: Fabp7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030832.2 | NP_110459.1 | 3 | 419 | ||
| GenBank mRNA | U02096.1 | - | 3 | 420 | ||