siRNA Details
| Gene Symbol: Grb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030846.2 | NP_110473.2 | 4 | 402 | ||
| GenBank mRNA | AA926364.1 | - | 4 | 402 | ||
| BC091144.1 | - | 3 | 346 | 7382476 | ||
| D49846.1 | - | 4 | 517 | |||
| FQ220840.1 | - | 4 | 468 | |||
| X62853.1 | - | 3 | 252 | |||

Grb2
Genomic Map