siRNA Details
| Gene Symbol: Nrp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030869.3 | NP_110496.1 | 2 | 374 | ||
| XM_006245039.2 | XP_006245101.1 | 2 | 932 | |||
| XM_006245040.2 | XP_006245102.1 | 2 | 931 | |||
| XM_006245041.2 | XP_006245103.1 | 2 | 933 | |||
| XM_006245042.2 | XP_006245104.1 | 2 | 932 | |||
| XM_008767137.1 | XP_008765359.1 | 2 | 931 | |||
| GenBank mRNA | AF016297.1 | - | 2 | 407 | ||

Nrp2
Genomic Map