siRNA Details
| Gene Symbol: Abat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031003.2 | NP_112265.1 | 3, 4 | 277 | ||
| XM_006245760.2 | XP_006245822.1 | 3, 4 | 485 | |||
| GenBank mRNA | BC081787.1 | - | 3, 4 | 274 | 7109775 | |
| D87839.1 | - | 3, 4 | 278 | |||
| U29701.1 | - | 2, 3 | 176 | |||

Abat
Genomic Map