siRNA Details
Gene Symbol: Gabbr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031028.3 | NP_112290.2 | 10, 11 | 1345 | ||
XM_006255879.2 | XP_006255941.1 | 10, 11 | 1439 | |||
XM_008772700.1 | XP_008770922.1 | 9, 10 | 1418 | |||
GenBank mRNA | AB016160.1 | - | 5, 6 | 888 | ||
AB016161.1 | - | 5, 6 | 888 | |||
AF283276.1 | - | 9, 10 | 1259 | |||
AJ831471.1 | - | 10, 11 | 1343 | |||
AJ831472.1 | - | 10, 11 | 1467 | |||
Y10369.2 | - | 10, 11 | 1317 | |||
Y10370.2 | - | 5, 6 | 990 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
