siRNA Details
| Gene Symbol: EPHB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004443.3 | NP_004434.2 | 10 | 2263 | ||
| GenBank mRNA | BC052968.1 | - | 10 | 2246 | 6472607 | |
| X75208.1 | - | 10 | 1853 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

EPHB3| Gene Symbol: EPHB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004443.3 | NP_004434.2 | 10 | 2263 | ||
| GenBank mRNA | BC052968.1 | - | 10 | 2246 | 6472607 | |
| X75208.1 | - | 10 | 1853 | |||