siRNA Details
| Gene Symbol: Grm7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031040.1 | NP_112302.1 | 1 | 638 | ||
| XM_008763199.1 | XP_008761421.1 | 1 | 638 | |||
| GenBank mRNA | D16817.1 | - | 1 | 638 | ||
| U06832.1 | - | 1 | 373 | |||
More Information
Other Species Targeted
Set Membership: |
|

Grm7
Genomic Map