siRNA Details
| Gene Symbol: Jup | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031047.2 | NP_112309.2 | 4 | 882 | ||
| XM_006247429.1 | XP_006247491.1 | 4 | 653 | |||
| XM_006247430.2 | XP_006247492.1 | 4 | 619 | |||
| GenBank mRNA | BC065580.1 | - | 4 | 882 | 5599724 | |
| FQ223166.1 | - | 4 | 642 | |||
| U58858.1 | - | 4 | 634 | |||

Jup
Genomic Map