siRNA Details
| Gene Symbol: Aldh6a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031057.2 | NP_112319.2 | 4 | 361 | ||
| XM_006240354.2 | XP_006240416.1 | 4 | 726 | |||
| XM_008764862.1 | XP_008763084.1 | 3 | 265 | |||
| XM_008764863.1 | XP_008763085.1 | 4 | 363 | |||
| GenBank mRNA | DN948946.1 | - | 4 | 358 | ||

Aldh6a1
Genomic Map