siRNA Details
| Gene Symbol: Msh2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031058.1 | NP_112320.1 | 1 | 192 | ||
| GenBank mRNA | BC161846.1 | - | 1 | 208 | 9029023 | |
| X93591.1 | - | 1 | 192 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Msh2| Gene Symbol: Msh2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031058.1 | NP_112320.1 | 1 | 192 | ||
| GenBank mRNA | BC161846.1 | - | 1 | 208 | 9029023 | |
| X93591.1 | - | 1 | 192 | |||