siRNA Details
| Gene Symbol: Bmpr1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030849.1 | NP_110476.1 | 3 | 392 | ||
| GenBank mRNA | D17667.1 | - | 2 | 167 | ||
| D38082.1 | - | 2 | 249 | |||
| S75359.1 | - | 3 | 392 | |||
| Gene Symbol: LOC686129 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008767400.1 | XP_008765622.1 | 1 | 403 | ||

Bmpr1a
Genomic Map