siRNA Details
Gene Symbol: Mia | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030852.2 | NP_110479.1 | 3 | 284 | ||
GenBank mRNA | U51438.1 | - | 3 | 174 | ||
U67884.1 | - | 3 | 284 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Mia | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030852.2 | NP_110479.1 | 3 | 284 | ||
GenBank mRNA | U51438.1 | - | 3 | 174 | ||
U67884.1 | - | 3 | 284 |