siRNA Details
| Gene Symbol: Nfkbib | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030867.2 | NP_110494.2 | 3 | 622 | ||
| GenBank mRNA | AF246634.1 | - | 3 | 445 | ||
| BC085729.1 | - | 3 | 622 | 7114624 | ||
| FQ228042.1 | - | 3 | 419 | |||
| FQ235103.1 | - | 3 | 567 | |||

Nfkbib
Genomic Map