siRNA Details
| Gene Symbol: Pld1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030992.1 | NP_112254.1 | 2 | 305 | ||
| XM_006232205.2 | XP_006232267.1 | 2 | 1080 | |||
| XM_006232206.2 | XP_006232268.1 | 3 | 733 | |||
| XM_008760891.1 | XP_008759113.1 | 3 | 733 | |||
| GenBank mRNA | AB000778.1 | - | 2 | 320 | ||
| AB000779.1 | - | 2 | 305 | |||
| AB003170.1 | - | 2 | 197 | |||
| AB003171.1 | - | 2 | 197 | |||
| AF017251.1 | - | 1 | 149 | |||
| U69550.1 | - | 2 | 305 | |||
| U88986.1 | - | 2 | 217 | |||

Pld1
Genomic Map