siRNA Details
| Gene Symbol: Amhr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030998.1 | NP_112260.1 | 2, 3 | 315 | ||
| GenBank mRNA | KJ923229.1 | - | 2, 3 | 315 | ||
| U42427.1 | - | 2, 3 | 315 | |||
| X71916.1 | - | 2, 3 | 297 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Amhr2| Gene Symbol: Amhr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030998.1 | NP_112260.1 | 2, 3 | 315 | ||
| GenBank mRNA | KJ923229.1 | - | 2, 3 | 315 | ||
| U42427.1 | - | 2, 3 | 315 | |||
| X71916.1 | - | 2, 3 | 297 | |||