siRNA Details
| Gene Symbol: Abat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031003.2 | NP_112265.1 | 8 | 569 | ||
| XM_006245760.2 | XP_006245822.1 | 8 | 777 | |||
| GenBank mRNA | BC081787.1 | - | 8 | 566 | 7109775 | |
| D87839.1 | - | 8 | 570 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Abat| Gene Symbol: Abat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031003.2 | NP_112265.1 | 8 | 569 | ||
| XM_006245760.2 | XP_006245822.1 | 8 | 777 | |||
| GenBank mRNA | BC081787.1 | - | 8 | 566 | 7109775 | |
| D87839.1 | - | 8 | 570 | |||